missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl ether, 99%

Product Code. 11412458
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412458 500 g 500g
11354479 250 g 250g
11422458 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412458 Supplier Thermo Scientific Alfa Aesar Supplier No. A15791.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 101-84-8 | C12H10O | 170.211 g/mol

Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

Solubility
Soluble in alcohol, chloroform, ethyl acetate, benzene, diethyl ether and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-84-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00003034
InChI Key USIUVYZYUHIAEV-UHFFFAOYSA-N
Synonym diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl
PubChem CID 7583
ChEBI CHEBI:39258
IUPAC Name phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2

Specifications

Melting Point 26°C to 29°C
Density 1.071
Boiling Point 258°C to 260°C
Flash Point 125°C (257°F)
Odor Aromatic
Refractive Index 1.5805
Quantity 500 g
UN Number UN3077
Beilstein 1364620
Merck Index 14,7288
Solubility Information Soluble in alcohol,chloroform,ethyl acetate,benzene,diethyl ether and glacial acetic acid. Insoluble in water.
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material Diphenyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.