missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphorochloridate, 97%

Product Code. 11490024
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490024 1000 g 1000g
11400034 50 g 50g
11410034 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490024 Supplier Thermo Scientific Alfa Aesar Supplier No. A13546.0B
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2524-64-3 | C12H10ClO3P | 268.633 g/mol

Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5',5'-triphosphate, guanosine-cytidine-5',5'-triphosphate, guanosine-uridine-5',5'-triphosphate, diguanosine-5',5'-diphosphate and diguanosine-5',5'-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5′,5′-triphosphate, guanosine-cytidine-5′,5′-triphosphate, guanosine-uridine-5′,5′-triphosphate, diguanosine-5′,5′-diphosphate and diguanosine-5′,5′-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.633
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Density 1.296
Boiling Point 145°C to 147°C (1 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.55
Quantity 1000 g
UN Number UN3265
Beilstein 654130
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 268.64
Percent Purity 97%
Chemical Name or Material Diphenyl phosphorochloridate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.