missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfone, 99+%

Product Code. 11348055
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348055 1000 g 1kg
11338055 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348055 Supplier Thermo Scientific Alfa Aesar Supplier No. L08232.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-63-9 | C12H10O2S | 218.27 g/mol

It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-63-9
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00007548
InChI Key KZTYYGOKRVBIMI-UHFFFAOYSA-N
Synonym diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
PubChem CID 31386
ChEBI CHEBI:78360
IUPAC Name benzenesulfonylbenzene
SMILES O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 123°C to 130°C
Density 1.36
Boiling Point 377°C to 379°C
Flash Point 184°C (363°F)
Odor Odorless
Quantity 1000 g
Beilstein 1910573
Merck Index 14,3238
Solubility Information Insoluble in water.
Formula Weight 218.27
Percent Purity ≥99%
Chemical Name or Material Diphenyl sulfone
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.