missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl terephthalate, 97%, Thermo Scientific™

Product Code. 11327885
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327885 25 g 25g
11317885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327885 Supplier Thermo Scientific Alfa Aesar Supplier No. L02877.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1539-04-4
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00016574
InChI Key HPGJOUYGWKFYQW-UHFFFAOYSA-N
Synonym diphenyl terephthalate, terephthalic acid, diphenyl ester, unii-s0kw8m264e, 1,4-benzenedicarboxylic acid, diphenyl ester, 1,4-benzenedicarboxylic acid, 1,4-diphenyl ester, 1,4-diphenyl benzene-1,4-dicarboxylate, diphenyl tere-phthalate, terephthalic acid diphenyl, acmc-1buw8, phenyl 4-phenoxycarbonyl benzoate
PubChem CID 73757
IUPAC Name diphenyl benzene-1,4-dicarboxylate
SMILES O=C(OC1=CC=CC=C1)C1=CC=C(C=C1)C(=O)OC1=CC=CC=C1

Specifications

Melting Point 196°C to 198°C
Odor Odorless
Quantity 25 g
Beilstein 1889204
Formula Weight 318.33
Percent Purity 97%
Chemical Name or Material Diphenyl terephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.