missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, 99%

Product Code. 11347695
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347695 500 g 500g
11337695 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347695 Supplier Thermo Scientific Alfa Aesar Supplier No. L02404.36
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-29-3 | C14H11N | 193.249 g/mol

Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-29-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00001862
InChI Key NEBPTMCRLHKPOB-UHFFFAOYSA-N
Synonym diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
PubChem CID 6837
IUPAC Name 2,2-diphenylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2

Specifications

Melting Point 72°C to 76°C
Boiling Point 181°C (12 mmHg)
Flash Point 120°C (248°F)
Quantity 500 g
Beilstein 1911160
Merck Index 14,3316
Solubility Information Insoluble in water.
Formula Weight 193.25
Percent Purity 99%
Chemical Name or Material Diphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.