missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, 99+%

Product Code. 11358511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11358511 2.5 kg Plastic bottle 2.5kg
10554081 5 g Glass bottle 5g
10225670 100 g Plastic bottle 100g
10665582 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11358511 Supplier Thermo Scientific Acros Supplier No. 150360025
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-29-3 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-29-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
InChI Key NEBPTMCRLHKPOB-UHFFFAOYSA-N
Synonym diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
PubChem CID 6837
IUPAC Name 2,2-diphenylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2

Specifications

Melting Point 73.0°C to 76.0°C
Color Cream to White or Yellow
Boiling Point 181.0°C (12.0 mmHg)
Flash Point 120°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)2CHCN
Quantity 2.5 kg
Beilstein 09,674
Merck Index 15,3346
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot alcohol, propylene glycol, hot, ligroin and ether, slightly soluble in cold ligroin
Formula Weight 193.25
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Diphenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.