missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, TRC

Product Code. 30136222
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136222 25 g 25g
30010892 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136222 Supplier Toronto Research Chemicals Supplier No. TRCD48945025G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-29-3
Molecular Formula C14 H11 N
Molecular Weight (g/mol) 193.24
Synonym Diphenylacetonitrile, Methadone Hydrochoride Imp. E (EP)
IUPAC Name 2,2-diphenylacetonitrile
SMILES N#CC(c1ccccc1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H11N/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H
Formula Weight 193.0891
Chemical Name or Material Diphenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.