missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride

Product Code. 15416609
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 15416609

Brand: Thermo Scientific Alfa Aesar J64838.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
MDL Number MFCD00214165
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Melting Point >310°C
Color White
Quantity 100 mg
Formula Weight 314.55
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.