accessibility menu, dialog, popup

UserName

ICl

CAS RN 4673-26-1

IUPAC Name: 8λ³-iodatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ylium chloride
Synonyms: 8λ³-iodatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ylium chloride
Molecular Weight (g/mol): 314.55
Molecular Formula: C12H8ClI
InChi Key: FCFZKAVCDNTYID-UHFFFAOYSA-M
SMILES: [Cl-].[I+]1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)