missing translation for 'onlineSavingsMsg'
Learn More

Diphenylglyoxime, 97%, Thermo Scientific™

Product Code. 10168112 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10168112

Brand: Thermo Scientific Acros 408930250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23873-81-6
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002113
InChI Key JJZONEUCDUQVGR-VCFJNTAESA-N
Synonym diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
PubChem CID 5369401
IUPAC Name N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
SMILES O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1

Specifications

Melting Point 238.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,1081
Solubility Information Solubility in water: insoluble.
Formula Weight 240.26
Percent Purity 97%
Grade Technical
Chemical Name or Material Diphenylglyoxime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.