missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium hexafluorophosphate, 98%

Product Code. 11499056
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499056 50 g 50g
11489056 10 g 10g
11409066 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499056 Supplier Thermo Scientific Alfa Aesar Supplier No. A17934.18
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58109-40-3 | C12H10I | 281.12 g/mol

Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyliodonium hexafluorophosphate is used as cationic photoinitiator and photoacid generator. It acts as a catalyst for the photochemical polymerization of various monomers. Further, it used as arylating reagent as well as an oxidant in organic synthesis.

Solubility
Soluble in tetrahydrofuran, ethyl acetate, methanol, acetonitrile, dimethylformamide, dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes, hexanes, toluene and diethyl ether.

Notes
Light sensitive and hygroscopic. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58109-40-3
Molecular Formula C12H10I
Molecular Weight (g/mol) 281.12
MDL Number MFCD00061398
InChI Key OZLBDYMWFAHSOQ-UHFFFAOYSA-N
Synonym diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
PubChem CID 2737136
IUPAC Name diphenyliodanium
SMILES [I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 139°C to 144°C
Quantity 50 g
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Soluble in tetrahydrofuran,ethyl acetate,methanol,acetonitrile,dimethylformamide,dimethyl carbonate. Sparingly soluble in chloroform and dichloromethane. Insoluble in pentanes,hexanes,toluene and diethyl ether.
Formula Weight 426.08
Percent Purity 98%
Chemical Name or Material Diphenyliodonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.