missing translation for 'onlineSavingsMsg'
Learn More

Dipropofol, TRC

Product Code. 30010030
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010030 250 mg 250mg
30008356 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010030 Supplier Toronto Research Chemicals Supplier No. TRCD492140250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2416-95-7
Molecular Formula C24 H34 O2
Molecular Weight (g/mol) 354.53
Synonym 3,3',5,5'-Tetrakis(1-methylethyl)biphenyl-4,4'-diol, 2,2',6,6'-Tetraisopropyl-p,p'-biphenol, 3,3',5,5'-Tetraisopropyl-4,4'-dihydroxybiphenyl, Dipropofol, Propofol Imp. E (EP)
IUPAC Name 4-[4-hydroxy-3,5-di(propan-2-yl)phenyl]-2,6-di(propan-2-yl)phenol
SMILES CC(C)c1cc(cc(C(C)C)c1O)c2cc(C(C)C)c(O)c(c2)C(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C24H34O2/c1-13(2)19-9-17(10-20(14(3)4)23(19)25)18-11-21(15(5)6)24(26)22(12-18)16(7)8/h9-16,25-26H,1-8H3
Formula Weight 354.2559
Percent Purity >95%
Chemical Name or Material Propofol Dimer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.