missing translation for 'onlineSavingsMsg'
Learn More

Diquat dibromide, Dr. Ehrenstorfer

Product Code. 30321671
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321671 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321671 missing translation for 'mfr' Dr. Ehrenstorfer missing translation for 'supplierNo' DRECA12960000100MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diquat dibromide
CAS 85-00-7
Analyte or Component Names Diquat dibromide
Shipping Condition +20°C
Molecular Formula C12 H12 N2 . 2 Br
Certifications/Compliance ISO 17034
Synonym Dipyrido[1,2-a:2',1'-c]pyrazinediium, 6,7-dihydro-, bromide (1:2), 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazidiinium dibromide (6CI), 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinediium dibromide (7CI), Dipyrido[1,2-a:2',1'-c]pyrazinediium, 6,7-dihydro-, dibromide (8CI,9CI), 1,1'-Dimethyl-4,4'-bipyridilium dibromide, 1,1'-Ethylene-2,2'-bipyridinium dibromide, 1,1'-Ethylene-2,2'-bipyridylium dibromide, 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinium dibromide, 9,10-Dihydro-8a,10a-diazoniaphenanthrene dibromide, Diquat, Diquat dibromide, Eliminator, Eliminator (herbicide), N,N'-Ethylene-2,2'-bipyridinium dibromide, Ortho-Diquat, PP 100, PP 100 (herbicide), Reglon, Reglone, Reglone Turbo, Reglox, Reward, Weedtrine D
InChI Formula InChI=1S/C12H12N2.2BrH/c1-3-7-13-9-10-14-8-4-2-6-12(14)11(13)5-1;;/h1-8H,9-10H2;2*1H/q+2;;/p-2
SMILES [Br-].[Br-].C1C[n+]2ccccc2c3cccc[n+]13
IUPAC Name 6,7-dihydrodipyrido[1,2-b:1',2'-e]pyrazine-5,8-diium;dibromide
Molecular Weight (g/mol) 344.05
Quantity 100 mg
Formula Weight 341.9367 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.