missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIS3L2 Antibody (6C7B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37316-0.025ml
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
DIS3L2 Monoclonal specifically detects DIS3L2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Tekniske data
| DIS3L2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| DIS3 mitotic control homolog (S. cerevisiae)-like 2, DIS3-like exonuclease 2, FAM6A, member A | |
| Mouse | |
| 99.3 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 6C7B2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DIS3L2 | |
| Purified recombinant fragment of human DIS3L2 (AA: 27-250) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 129563 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion