missing translation for 'onlineSavingsMsg'
Learn More

Disodium terephthalate, 99+%

Product Code. p-7058648
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11330218

Brand: Thermo Scientific Alfa Aesar 042946.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10028-70-3 | C8H4Na2O4 | 210.096 g/mol

Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-70-3
Molecular Formula C8H4Na2O4
Molecular Weight (g/mol) 210.096
MDL Number MFCD00013137
InChI Key VIQSRHWJEKERKR-UHFFFAOYSA-L
Synonym disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:?
PubChem CID 82305
IUPAC Name disodium;terephthalate
SMILES C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 210.1
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Disodium terephthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.