Learn More
Skytec™ Disposable Protective Tear-Off For Alpha Sentinel Full Face Mask
Product Code. 30242966 Shop All Skytec Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
30242966 10/Pk. Pack of 10
1 options
Product Code. 30242966 Supplier Skytec™ Supplier No. ASRAS0036BZ
  • ONLINE EXCLUSIVE PRICE
    32.51 €
    21.65 € / Pack of 10
    Save 10.86 €
    33% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Alpha Sentinel Protective Tear off protects the Sentinel Full Face Mask against damage from exposure to hazardous substances.
  • Replaceable PET film with two adhesive strips which affix it to the mask.
  • Tabs on each side of the tear off enable easy removal so that it can be replaced
  • A location feature central to the tear off is used to easily align it with two small ribs within the Alpha Sentinel Full Face Mask.
Specifications
Color Clear
Disposable Single-use
Filter Type Combination
Material Plastic
Product Type Protective Tear-Off
Size One Size
Facepiece Type Alpha Sentinel Full Face Mask
Quantity 10/Pk.