missing translation for 'onlineSavingsMsg'
Learn More

Dithiobiuret, 97%

Product Code. 11401980
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401980 5 g 5g
11411980 25 g 25g
11421980 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11401980 Supplier Thermo Scientific Alfa Aesar Supplier No. A16175.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-53-7 | C2H5N3S2 | 135.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-53-7
Molecular Formula C2H5N3S2
Molecular Weight (g/mol) 135.20
MDL Number MFCD00037832
InChI Key JIRRNZWTWJGJCT-UHFFFAOYSA-N
Synonym dithiobiuret, 2,4-dithiobiuret, thioimidodicarbonic diamide, biuret, dithio, imidodicarbonodithioic diamide, biuret, 2,4-dithio, imidodicarbonimidothioic diamide, rcra waste number p049, urea, 2-thio-1-thiocarbamoyl, allophanimidic acid, dithio
PubChem CID 2758725
IUPAC Name carbamothioylthiourea
SMILES NC(=S)NC(N)=S

Specifications

Melting Point ∼183°C (decomposition)
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 1756621
Merck Index 14,3371
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material Dithiobiuret

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.