missing translation for 'onlineSavingsMsg'
Learn More

Dithizone, TRC

Product Code. 30136216
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136216 10 g 10g
30012517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136216 Supplier Toronto Research Chemicals Supplier No. TRCD49429510G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13 H12 N4 S
Molecular Weight (g/mol) 256.33
Synonym (Phenylazo)thioformic Acid 2-Phenylhydrazide, DIZO, N,2-Diphenyldiazenecarbohydrazonothioic Acid, Diphenylthiocarbazone, Dithizon, Dithizone, NSC 215189, NSC 4275, NSC 97262, NSC 97264, NSC 97353, NSC 99124;
IUPAC Name (3E)-1-anilino-3-phenyliminothiourea
SMILES S=C(NNc1ccccc1)N=Nc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H12N4S/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)/b17-15+
Formula Weight 256.0783
Chemical Name or Material Dithizone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.