missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dityrosine Antibody (10A6), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59361MFV610
This item is not returnable.
View return policy
Description
Dityrosine Monoclonal antibody specifically detects Dityrosine in Non-species specific samples. It is validated for Flow Cytometry
Specifications
| Dityrosine | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Flow Cytometry | |
| 10A6 | |
| Flow Cytometry | |
| Dityrosine;Dityrosine (DT);DT;DY | |
| Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction