missing translation for 'onlineSavingsMsg'
Learn More

Divarinic Acid, TRC

Product Code. 30004100
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004100 25 mg 25mg
30009139 5 mg 5mg
30006564 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004100 Supplier Toronto Research Chemicals Supplier No. TRCD49446325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4707-50-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
Synonym 6-Propyl-b-resorcylic Acid, (7CI,8CI), 2,4-Dihydroxy-6-propylbenzoic Acid, Divaric Acid
SMILES OC(C(C(O)=CC(O)=C1)=C1CCC)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H12O4/c1-2-3-6-4-7(11)5-8(12)9(6)10(13)14/h4-5,11-12H,2-3H2,1H3,(H,13,14)
Formula Weight 196.0736
Percent Purity >95%
Chemical Name or Material Divarinic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.