Learn More
Invitrogen™ DJ-1 Monoclonal Antibody (4H4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA533099
Product Code. 16565731
  • 566.00 € / 100µL
Description
DJ-1 Monoclonal Antibody for Western Blot, ICC/IF, IHC
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DJ-1
Monoclonal
1 mg/mL
PBS with 10mM sodium azide
O88767, Q5E946, Q99497, Q99LX0
Park7
Full length recombinant human Park7 expressed in and purified from E. coli.
100 μL
Primary
Human, Mouse, Rat, Bovine
Antibody
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry
4H4
Unconjugated
Park7
CAP1; Contraception-associated protein 1; CTA-215D11.1; dj1; DJ-1; DJ-1 protein; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; HEL-S-67p; hypothetical protein LOC449674; maillard deglycase; Oncogene DJ1; OTTHUMP00000001350; OTTHUMP00000001351; PARK7; PARK7 GENE; PARK7/DJ1; parkinson disease (autosomal recessive, early onset) 7; Parkinson disease 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; Parkinson disease protein 7 homolog; protein/nucleic acid deglycase DJ-1; parkinson protein 7; Parkinsonism associated deglycase; parkinsonism-associated deglycase; Protein CAP1; protein deglycase DJ-1; protein deglycase DJ-1zDJ-1; Protein DJ-1; protein DJ-1-like protein; protein DJ-1zDJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; SP22; zDJ-1; zgc:103725
Mouse
Protein G
RUO
11315, 117287, 511268, 57320
-20°C, Avoid Freeze/Thaw Cycles