missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 453.00 €
Specifications
| Antigen | Dkk-1 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 0.5-2.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18128801
|
Novus Biologicals
NBP2-24701SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790514
|
Novus Biologicals
NBP2-24701 |
0.1 mg |
453.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Dkk-1 | |
| Western Blot 0.5-2.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| DKK1 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, 97% homologous in mouse, dog and horse, 95% homologous in rat and 94% homologous in cow. |
| 1.0 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 22943 | |
| A portion of amino acids 150-200 of human DKK1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title