missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-1 Rabbit anti-Human, Clone: R015, Novus Biologicals™
Rabbit Monoclonal Antibody
506.00 € - 673.00 €
Specifications
| Antigen | Dkk-1 |
|---|---|
| Clone | R015 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214717
|
Novus Biologicals
NBP2-89310-200UG |
200 μg |
506.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18277887
|
Novus Biologicals
NBP2-89310-500UG |
500 μg |
673.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Neutralization.Specifications
| Dkk-1 | |
| Neutralization | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
| 22943 | |
| IgG | |
| Protein A purified |
| R015 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| Recombinant Human Dkk-1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title