missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-3 Antibody (8A5C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 € - 557.00 €
Specifications
| Antigen | Dkk-3 |
|---|---|
| Clone | 8A5C6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698705
|
Novus Biologicals
NBP2-37430-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18167618
|
Novus Biologicals
NBP2-37430 |
0.1 mL |
557.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Dkk-3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| DKK3 | |
| IgG1 | |
| Affinity Purified |
| 8A5C6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 27122 | |
| Purified recombinant fragment of human DKK3 (AA: 91-350 ) expressed in E. Coli. | |
| Primary | |
| 38.3 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title