missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-2-Benzylserine, 97%

Product Code. 10420052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420052 1 g Glass bottle 1g
10736623 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420052 Supplier Thermo Scientific Chemicals Supplier No. 339780010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4740-47-0 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4740-47-0
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD01863305
InChI Key ZMNNAJIBOJDHAF-JTQLQIEISA-N
Synonym dl-2-benzylserine, 2-amino-3-hydroxy-2-benzylpropanoic acid, alpha-benzyl-dl-serine
PubChem CID 7010088
IUPAC Name (2S)-2-amino-2-benzyl-3-hydroxypropanoic acid
SMILES C1=CC=C(C=C1)CC(CO)(C(=O)O)N

Specifications

Melting Point 245°C to 263°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water
Formula Weight 195.22
Percent Purity 97%
Physical Form Powder or Flakes
Chemical Name or Material DL-2-Benzylserine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Benzylserine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.