missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Pentanol, 98%

Product Code. 10467710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10467710 500 mL Glass bottle 500mL
10397150 1 L Glass bottle 1 L
10616724 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10467710 Supplier Thermo Scientific Acros Supplier No. 129985000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6032-29-7 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6032-29-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004579
InChI Key JYVLIDXNZAXMDK-UHFFFAOYSA-N
Synonym 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
PubChem CID 22386
ChEBI CHEBI:77518
IUPAC Name pentan-2-ol
SMILES CCCC(C)O

Specifications

Melting Point -50°C
Color Colorless
Density 0.8120g/mL
Boiling Point 118°C to 119°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(OH)CH3
Refractive Index 1.4050 to 1.407
Quantity 500 mL
Beilstein 01, 384
Residue after Evaporation 0.003% max.
Merck Index 15, 7231
Specific Gravity 0.812
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with alcohol and ether
Viscosity 4.1 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DL-2-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.