missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylpropionic acid, 98%

Product Code. 10001320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10001320 25 g Glass bottle 25g
10745431 10 g Glass bottle 10g
10666734 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10001320 Supplier Thermo Scientific Acros Supplier No. 130870250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 492-37-5 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-37-5
MDL Number MFCD00002650
InChI Key YPGCWEMNNLXISK-UHFFFAOYSA-N
Synonym 2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
PubChem CID 10296
ChEBI CHEBI:48526
IUPAC Name 2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Color Yellow
Density 1.1g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3CH(C6H5)CO2H
Refractive Index 1.5215 to 1.5235 (20°C, 589nm)
Quantity 25 g
Beilstein 09, 524
Molecular Weight (g/mol) 150.177
Formula Weight 150.18
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Phenylpropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.