missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Phenyllactic acid, 97%

Product Code. 10546830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10546830 10 g Glass bottle 10g
10017859 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10546830 Supplier Thermo Scientific Acros Supplier No. 427770100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 828-01-3 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-01-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00065928
InChI Key VOXXWSYKYCBWHO-UHFFFAOYSA-N
Synonym dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
PubChem CID 3848
ChEBI CHEBI:25998
IUPAC Name 2-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)O

Specifications

Melting Point 95°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(OH)COOH
Quantity 10 g
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material DL-3-Phenyllactic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.