Learn More
(1-Hexyl)triphenylphosphonium bromide, 98%
Description
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
Specifications
Specifications
| Melting Point | 199°C to 202°C |
| Quantity | 25 g |
| Beilstein | 3580731 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water and methanol. |
| IUPAC Name | hexyltriphenylphosphanium bromide |
| Formula Weight | 427.37 |
| Percent Purity | 98% |
| Chemical Name or Material | (1-Hexyl)triphenylphosphonium bromide |
Safety and Handling
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Hexyltriphenylphosphonium bromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.