missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Alanine, 99%

Product Code. 11474913
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474913 5000 g 5000g
11464913 250 g 250g
11454913 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474913 Supplier Thermo Scientific Chemicals Supplier No. A12230.0I
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol

DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
Synonym dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
PubChem CID 602
ChEBI CHEBI:16449
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O

Specifications

Melting Point ∼292°C (decomposition)
Beilstein 635807
Merck Index 14,204
Solubility Information Soluble in water. Slightly soluble in alcohol.
Density 1.42
Quantity 5000 g
Formula Weight 89.09
Percent Purity 99%
Chemical Name or Material DL-Alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.