missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine, 96%

Product Code. 15482558
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482558 5 g 5g
15492558 25 g 25g
15402568 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482558 Supplier Thermo Scientific Alfa Aesar Supplier No. H56126.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3374-22-9 | C3H7NO2S | 121.15 g/mol

DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3374-22-9
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00004881
InChI Key XUJNEKJLAYXESH-UHFFFAOYNA-N
Synonym dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
PubChem CID 594
ChEBI CHEBI:15356
IUPAC Name 2-amino-3-sulfanylpropanoic acid
SMILES NC(CS)C(O)=O

Specifications

Melting Point 225°C (decomposition)
Beilstein 1721406
Solubility Information Soluble in water (partly),1 N HCl (25mg/ml),ethanol,and acetic acid. Insoluble in acetone.
Quantity 5 g
Formula Weight 121.16
Percent Purity 96%
Chemical Name or Material DL-Cysteine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.