missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoproterenol hydrochloride, 98%

Product Code. 15404309
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404309 5 g 5g
15414309 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404309 Supplier Thermo Scientific Alfa Aesar Supplier No. J61788.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard selective beta-adrenoceptor agonist | CAS: 51-30-9 | C11H18ClNO3 | 247.72 g/mol

Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-30-9
Molecular Formula C11H18ClNO3
Molecular Weight (g/mol) 247.72
MDL Number MFCD00012603,MFCD00064548
InChI Key IROWCYIEJAOFOW-UHFFFAOYNA-N
Synonym isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
PubChem CID 5807
IUPAC Name hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 165°C to 175°C (decomposition)
Color White to Yellow
Quantity 5 g
Beilstein 3917363
Merck Index 14,5217
Solubility Information Soluble in water (50mg/ml at 25°C),ethanol (20mg/ml),methanol,DMSO (50mg/ml at 25°C),and phosphate buffered saline (50 mM).
Formula Weight 247.72
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Isoproterenol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.