missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoproterenol hydrochloride, 98%, Thermo Scientific™

Standard selective beta-adrenoceptor agonist

Brand:  Thermo Scientific Alfa Aesar J61788.14

Quantity 25g

 View more versions of this product

Product Code. 15414309

  • 217.00 € / 25g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

51-30-9
247.72
IROWCYIEJAOFOW-UHFFFAOYNA-N
5807
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
C11H18ClNO3
MFCD00012603,MFCD00064548
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride

Specifications

51-30-9
MFCD00012603,MFCD00064548
3917363
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
IROWCYIEJAOFOW-UHFFFAOYNA-N
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
5807
98%
C11H18ClNO3
25g
14,5217
Soluble in water (50mg/ml at 25°C),ethanol (20mg/ml),methanol,DMSO (50mg/ml at 25°C),and phosphate buffered saline (50 mM).
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
247.72
247.72
DL-Isoproterenol hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.