missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrate, 98+%

Product Code. 11361957
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361957 25 g 25g
11351957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361957 Supplier Thermo Scientific Alfa Aesar Supplier No. L07576.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 885701-25-7 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885701-25-7
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151036
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
Synonym dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate
PubChem CID 2734605
IUPAC Name 2,6-diaminohexanoic acid;hydrate
SMILES O.NCCCCC(N)C(O)=O

Specifications

Melting Point ∼110°C
Beilstein 1616991
Quantity 25 g
Formula Weight 164.21 (146.19 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material DL-Lysine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.