accessibility menu, dialog, popup

UserName

CAS RN 885701-25-7

H2NOHNH2OH2O

CAS RN 885701-25-7

IUPAC Name: 2,6-diaminohexanoic acid hydrate
Synonyms: lysine hydrate
Molecular Weight (g/mol): 164.21
Molecular Formula: C6H16N2O3
InChi Key: HZRUTVAFDWTKGD-UHFFFAOYNA-N
SMILES: O.NCCCCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)