missing translation for 'onlineSavingsMsg'
Learn More

DL-O-Phosphoserine, MedChemExpress

Product Code. 16496636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16496636 Supplier Medchem Express Supplier No. HY15130

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress DL-O-Phosphoserine, a normal metabolite in human biofluid, is an ester of serine and phosphoric acid.

DL-O-Phosphoserine, a normal metabolite in human biofluid, is an ester of serine and phosphoric acid.

Specifications

Chemical Name or Material DL-O-Phosphoserine
Physical Form Solid
CAS 17885-08-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H8NO6P
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : 25 mg/mL (135.08 mM; Need ultrasonic)
SMILES OP(OCC(C(O)=O)N)(O)=O
Molecular Weight (g/mol) 185.07
Formula Weight 185.07
Percent Purity 98.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.