missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLEC/CLEC4C/BDCA-2 Antibody (3F9B1), Novus Biologicals™
Mouse Monoclonal Antibody
298.00 € - 511.00 €
Specifications
| Antigen | DLEC/CLEC4C/BDCA-2 |
|---|---|
| Clone | 3F9B1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30033665
|
Novus Biologicals™
NBP3-27163-0.1ml |
0.1 mL |
511.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028647
|
Novus Biologicals™
NBP3-27163-0.025ml |
0.025 mL |
298.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| DLEC/CLEC4C/BDCA-2 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Dendritic Cell Markers | |
| PBS | |
| 170482 | |
| IgG1 | |
| Protein G purified |
| 3F9B1 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| Purified recombinant fragment of human DLEC/CLEC4C/BDCA-2 (AA: extra 45-213) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title