missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLG7/HURP Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB110-40572PCP
This item is not returnable.
View return policy
Description
DLG7/HURP Polyclonal antibody specifically detects DLG7/HURP in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| DLG7/HURP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| PerCP | |
| DAP-5, Discs large homolog 7, discs, large (Drosophila) homolog-associated protein 5, disks large-associated protein 5, DLG7, Hepatoma up-regulated protein, HURPDLG1, KIAA0008discs, large homolog 7 (Drosophila), large homolog 7 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of Human Hepatoma Upregulated Protein/Discs, Large 7 using the numbering given in entry NP_055565.2 (GeneID 9787). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 9787 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction