missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL3 Antibody (DLL3/9361), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44583-100ug
This item is not returnable.
View return policy
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Immunohistochemistry
Specifications
| DLL3 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| DLL3/9361 | |
| Unconjugated | |
| delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
| Recombinant fragment (around aa400-618) of human DLL3 protein (Uniprot: Q9NYJ7). Exact sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Neuroscience | |
| 10683 | |
| Store at 4⪚C. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction