missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLL3 Antibody (Dragonfly patent anti-DLL3) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00 € - 920.00 €
Specifications
| Antigen | DLL3 |
|---|---|
| Clone | Dragonfly patent anti-DLL3 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047787
|
Novus Biologicals™
NBP3-28318-50ug |
50 μg |
213.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046222
|
Novus Biologicals™
NBP3-28318-1mg |
1 mg |
920.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044721
|
Novus Biologicals™
NBP3-28318-100ug |
100 μg |
338.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| DLL3 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cell Biology, Cell Cycle and Replication, Neuroscience | |
| Q9NYJ7 | |
| 10683 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Dragonfly patent anti-DLL3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3 | |
| DLL3 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title