missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLL4 Antibody (navicixizumab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28485-1mg
This item is not returnable.
View return policy
Description
DLL4 Monoclonal antibody specifically detects DLL4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DLL4 | |
| Monoclonal | |
| Unconjugated | |
| Q9NR61 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| navicixizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
| DLL4 | |
| 1 mg | |
| Neuroscience | |
| 54567 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction