missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM/F-12, powder, HEPES
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components.
Product Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM/F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DMEM/F-12 product made in our Grand Island facility (12400-024). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM /F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation (see protocol for details).
Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components.
Product Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM/F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DMEM/F-12 product made in our Grand Island facility (12400-024). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM /F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications
Specifications
| Form | Powder |
| Product Type | Dulbecco's Modified Eagle Medium: Nutrient Mixture F-12 |
| With Additives | Glutamine, HEPES, Phenol Red |
| Without Additives | No Sodium Bicarbonate |
| For Use With (Application) | Mammalian cell culture |
| Quantity | 10 L |
| Certifications/Compliance | ISO 13485 standards, FDA |
| Content And Storage | 2 to 8°C |
| Description | Dulbecco's Modified Eagle Medium: Nutrient Mixture F-12 (DMEM / F-12), 10L, Powder, HEPES Buffer, With L-Glutamine, HEPES, Phenol Red, 12 Months Shelf Life, For Mammalian Cell Culture |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?