missing translation for 'onlineSavingsMsg'
Learn More

DMSA 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318025
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318025 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318025 Supplier Dr. Ehrenstorfer Supplier No. DREA13030000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DMSA
CAS 4710-17-2
Analyte or Component Names DMSA
Shipping Condition Room Temperature
Molecular Formula C8 H12 N2 O2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C8H12N2O2S/c1-10(2)13(11,12)9-8-6-4-3-5-7-8/h3-7,9H,1-2H3
SMILES CN(C)S(=O)(=O)Nc1ccccc1
IUPAC Name (dimethylsulfamoylamino)benzene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 200.26
Quantity 1 mL
Formula Weight 200.0619 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.