missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase IV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-87405-25ul
This item is not returnable.
View return policy
Description
DNA Ligase IV Polyclonal specifically detects DNA Ligase IV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DNA Ligase IV | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| DNA joinase, DNA ligase 4, DNA ligase IV, DNA repair enzyme, EC 6.5.1.1, ligase IV, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 4, polynucleotide ligase, sealase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| LIG4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TYCVIAGSENIRVKNIILSNKHDVVKPAWLLECFKTKSFVPWQPRFMIHMCPSTKEHFAREYDCYGDSYFIDTDLNQLKEVFSGIKNSNEQTPEEMASLIADLEYRYSWDCSPLSMFRRHTVYLDSYA | |
| 25 μL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
| 3981 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction