missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05939-100ul
This item is not returnable.
View return policy
Description
DNA Polymerase beta Polyclonal antibody specifically detects DNA Polymerase beta in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| DNA Polymerase beta | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| DNA pol beta, DNA polymerase beta, DNA polymerase beta subunit, EC 2.7.7.7, EC 4.2.99.-, MGC125976, polymerase (DNA directed), beta | |
| Produced in rabbits immunized with E. coli-derived Human DNA Polymerase beta fragment. | |
| 100 μg | |
| DNA Polymerases, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 5423 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction