Learn More
DNA Polymerase gamma Antibody (S06-1G0), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19996-100ul
 View more versions of this product
Product Code. 18644154
  • 645.00 € / 100µL
Description
DNA Polymerase gamma Monoclonal antibody specifically detects DNA Polymerase gamma in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA Polymerase gamma
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
S06-1G0
Unconjugated
DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE
A synthetic peptide of human DNA Polymerase gamma (Uniprot # P54098)
100 μg
DNA Polymerases, DNA Repair
5428
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.