Learn More
DNA Polymerase gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-17832-100UL
 View more versions of this product
Product Code. 18369726
  • 703.00 € / 100µL
Description
DNA Polymerase gamma Polyclonal antibody specifically detects DNA Polymerase gamma in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA Polymerase gamma
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE
This antibody was developed against Recombinant Protein corresponding to amino acids: PKLMALTWDGFPLHYSERHGWGYLVPGRRDNLAKLPTGTTLESAGVVCPYRAIESLYRKHCLEQGKQQLMPQEAGLAEEFLLTD
100 μg
DNA Polymerases, DNA Repair
5428
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified