missing translation for 'onlineSavingsMsg'
Learn More

DNA Polymerase iota Antibody (OTI5A4), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

256.00 € - 550.00 €

Specifications

Antigen DNA Polymerase iota
Clone OTI5A4
Dilution Western Blot 1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306532
View Documents
Novus Biologicals
NBP3-44871-30ul
30 μL
256.00 €
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306551
View Documents
Novus Biologicals
NBP3-44871-100ul
100 μL
550.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNA Polymerase iota Monoclonal antibody specifically detects DNA Polymerase iota in Human samples. It is validated for Western Blot
Specifications

Specifications

DNA Polymerase iota
Western Blot 1:2000
Monoclonal
Purified
RUO
Human
DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB
Human recombinant protein fragment corresponding to amino acids 458-740 of human POLI (NP_009126) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI5A4
Western Blot
Unconjugated
Mouse
Chromatin Research, DNA Polymerases, DNA Repair
PBS (pH 7.3) 1% BSA, 50% glycerol
11201
IgG1
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.