Learn More
DNA-RNA Hybrid Antibody (S9.6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44628-20ug
 View more versions of this product
Product Code. 30303464
  • 256.00 € / 20µg
Description
DNA-RNA Hybrid Monoclonal antibody specifically detects DNA-RNA Hybrid in All samples. It is validated for ChIP assay,Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA-RNA Hybrid
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase
20 μg
Primary
Store at 4⪚C.
IgG2a κ
ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence
S9.6
Unconjugated
Mouse
Protein A purified
RUO
All species
Purified