missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC30 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00 € - 322.00 €
Specifications
| Antigen | DNAJC30 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640707
|
Novus Biologicals
NBP3-12538-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615367
|
Novus Biologicals
NBP3-12538-50ul |
50 μg |
183.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJC30 Polyclonal antibody specifically detects DNAJC30 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| DNAJC30 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DnaJ (Hsp40) homolog, subfamily C, member 30, dnaJ homolog subfamily C member 30, MGC12943, WBSCR18, Williams Beuren syndrome chromosome region 18, Williams-Beuren syndrome chromosomal region 18 protein | |
| Produced in rabbits immunized with purified, recombinant Human DNAJC30 (Uniprot#: Q96LL9; Ser39-Gly124) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 84277 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title